Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Suture, Size 2-0, No Needle, 54in, Violet, 36/Box
Catalog No. 502522351
Encompass_Preferred
The Ethicon J615H suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 54in in length, has a size of 2-0, and are dyed violet to provide greater visibility while suturing. This suture has no needle and has 36 sutures in a box.
Catalog No. 50-252-2351 Supplier Med Vet International Supplier No. J615H
The Ethicon J615H suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 54in in length, has a size of 2-0, and are dyed violet to provide greater visibility while suturing. This suture has no needle and has 36 sutures in a box.