Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Suture, Size 2, TP-1, 27in, Violet, 12/Box
Catalog No. 502522350
Encompass_Preferred
The Ethicon J649G suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 27in in length, has a size of 2, and are dyed violet to provide greater visibility while suturing. This suture comes with a TP-1 65mm 1/2c Taper Needle and has 12 sutures in a box.
Catalog No. 50-252-2350 Supplier Med Vet International Supplier No. J649G
The Ethicon J649G suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 27in in length, has a size of 2, and are dyed violet to provide greater visibility while suturing. This suture comes with a TP-1 65mm 1/2c Taper Needle and has 12 sutures in a box.