Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Plus Antibacterial Suture, Size 3-0, CT-3, 27in, Violet, 36/Box
Catalog No. 502522156
Encompass_Preferred
The Ethicon VCP327H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 27in in length, has a size of 3-0, and are dyed violet to provide greater visibility while suturing. This suture comes with a CT-3 22mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-252-2156 Supplier Med Vet International Supplier No. VCP327H
The Ethicon VCP327H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 27in in length, has a size of 3-0, and are dyed violet to provide greater visibility while suturing. This suture comes with a CT-3 22mm 1/2c Taper Needle and has 36 sutures in a box.