Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Suture, Size 5-0, TF, 27in, Undyed, 36/Box
Catalog No. 502522358
Encompass_Preferred
The Ethicon J433H suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 27in in length, has a size of 5-0, and are undyed. This suture comes with a TF 13mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-252-2358 Supplier Med Vet International Supplier No. J433H
The Ethicon J433H suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 27in in length, has a size of 5-0, and are undyed. This suture comes with a TF 13mm 1/2c Taper Needle and has 36 sutures in a box.