Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Suture, Size 6-0, PC-1, 18in, Undyed, 12/Box
Catalog No. 502522348
Encompass_Preferred
The Ethicon J833G suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 18in in length, has a size of 6-0, and are undyed. This suture comes with a PC-1 13mm 3/8c Conventional Cutting Needle and has 12 sutures in a box.
Catalog No. 50-252-2348 Supplier Med Vet International Supplier No. J833G
The Ethicon J833G suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 18in in length, has a size of 6-0, and are undyed. This suture comes with a PC-1 13mm 3/8c Conventional Cutting Needle and has 12 sutures in a box.