Learn More
Med Vet International Ethicon Vicryl Rapide Suture, Size 3-0, PS-2, 18in, 12/Box
Catalog No. 502522144
Encompass_Preferred
The Ethicon VR497 suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 3-0, they are dyed black to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a PS-2 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-252-2144 Supplier Med Vet International Supplier No. VR497
The Ethicon VR497 suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 3-0, they are dyed black to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a PS-2 Precision Point reverse cutting needle and has 12 sutures in a box.