Learn More
Med Vet International Ethicon Vicryl Rapide, 5-0 Polyglactin Suture, 18in P-3, 12/Box
Catalog No. 502522146
Encompass_Preferred
The Ethicon VR493 suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 5-0, they are dyed black to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-252-2146 Supplier Med Vet International Supplier No. VR493
The Ethicon VR493 suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 5-0, they are dyed black to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.