Learn More
Med Vet International Ethicon Ethilon Nylon Suture, Size 0, TP-1, 96In, Black, 12/Box
Supplier:  Med Vet International L880G
Encompass_Preferred
The Ethicon L880G suture is a high-quality non-absorbable Ethilon Nylon suture. It measures 96in in length, has a size of 0, and are dyed black to provide greater visibility while suturing. This suture comes with a TP-1 65mm 1/2c Taper Needle and has 12 sutures in a box.
Catalog No. 50-255-9563