Learn More
Med Vet International Ethicon Ethilon Nylon Suture, Size 4-0, P-3, 18In, Black, 36/Box
Supplier:  Med Vet International 699H
Encompass_Preferred
The Ethicon 699H suture is a high-quality non-absorbable Ethilon Nylon suture. It measures 18in in length, has a size of 4-0, and are dyed black to provide greater visibility while suturing. This suture comes with a P-3 13mm 3/8c Reverse Prime Cutting Needle and has 36 sutures in a box.
Catalog No. 50-255-9699