Learn More
Med Vet International Ethicon Monocryl Plus Antibacterial Suture, Size 5-0, P-3, 18In, Violet, 12/Box
Supplier:  Med Vet International MCP463G
Encompass_Preferred
The Ethicon MCP214H suture is a high-quality absorbable Monocryl Poliglecaprone 25 suture. It measures 18in in length, has a size of 5-0, and are dyed violet to provide greater visibility while suturing. This suture comes with a P-3 13mm 3/8c Reverse Prime Cutting Needle and has 12 sutures in a box.
Catalog No. 50-255-9553