Learn More
Med Vet International Oasis Monofyl (Poliglecaprone) Suture, Size 0, NCT-1 Needle, 30in, 12/Box
Supplier:  Med Vet International MVY346
Encompass_Preferred
The Oasis Monofyl suture with needle is a monofilament synthetic absorbable suture that is comparable to Ethicon™ Monocryl®. The Monofyl suture has an absorption rate between 91 and 119 days. Size: 0. Needle Type: NCT-1, 36mm 1/2 Circle Taper Point. Length: 30in. Quantity: 12/box
Catalog No. 50-255-9360