Learn More
Med Vet International Oasis Monofyl (Poliglecaprone) Suture, Size 3-0, NFS-2 Needle, 30in, 12/Box
Supplier:  Med Vet International MVY923
Encompass_Preferred
The Oasis Monofyl suture with needle is a monofilament synthetic absorbable suture that is comparable to Ethicon™ Monocryl®. The Monofyl suture has an absorption rate between 91 and 119 days. Size: 3-0. Needle Type: NFS-2, 19mm 3/8 Circle Reverse Cutting. Length: 30in. Quantity: 12/box
Catalog No. 50-255-9358
 Restricted Item