Learn More
Med Vet International Oasis Nylon Suture, Size 1, with NFSL Needle, 12/box, Veterinary Use Only
Supplier:  Med Vet International MVN666V
Encompass_Preferred
The veterinary use Oasis Nylon suture with needle is a monofilament non-absorbable suture that is comparable to Ethicon™ Ethilon®. This Nylon suture has extremely high tensile strength that exceeds USP requirements. It provides excellent knot security, while the smooth surface allows for easy passage through the tissue. The thread color of this suture is blue and has a composition of polyamide. Size: 1. Needle Type: NFSL, 30mm 3/8 Circle Reverse Cutting. Length: 30in. Quantity: 12/box
Catalog No. 50-255-9365
 Restricted Item