Learn More
Med Vet International Oasis PDO Suture, Size 1, with NCP-1 Needle, 12/box
Supplier:  Med Vet International MVZ468
Encompass_Preferred
The Oasis PDO suture with needle is a monofilament synthetic absorbable suture that is comparable to Ethicon™ PDS®. The PDO smooth monofilament suture has a gradual absorption rate between 180 and 220 days. It retains 50% of its tensile strength for up to 6 weeks. This suture provides excellent pliability and holding strength. Size: 1. Needle Type: NCP-1, 36mm 1/2 Circle Reverse Cutting Point. Length: 30in. Quantity: 12/box
Catalog No. 50-255-9333