Learn More
Med Vet International Ethicon PDS II Suture, Size 1, CP-1, 27in, 36/Box
Catalog No. 502092843
Encompass_Preferred
The Ethicon Z468H suture is a high-quality absorbable Polydioxanone surgical suture. It measures 27in. in length, has a size of 1, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a CP-1 reverse cutting needle and has 36 sutures in a box.
Catalog No. 50-209-2843 Supplier Med Vet International Supplier No. Z468H
The Ethicon Z468H suture is a high-quality absorbable Polydioxanone surgical suture. It measures 27in. in length, has a size of 1, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a CP-1 reverse cutting needle and has 36 sutures in a box.