Learn More
Med Vet International Ethicon PDS II, 3-0 Polydioxanone Suture, 27in, FS-1, 36/Box
Catalog No. 502092851
Encompass_Preferred
The Ethicon Z442H suture is a high-quality absorbable Polydioxanone surgical suture. It measures 27in. in length, has a size of 3-0, they are dyed undyed to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a FS-1 reverse cutting needle and has 36 sutures in a box.
Catalog No. 50-209-2851 Supplier Med Vet International Supplier No. Z442H
The Ethicon Z442H suture is a high-quality absorbable Polydioxanone surgical suture. It measures 27in. in length, has a size of 3-0, they are dyed undyed to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a FS-1 reverse cutting needle and has 36 sutures in a box.