Learn More
Med Vet International Oasis PGA Suture, Size 2-0, with NSH Needle, 12/box
Supplier:  Med Vet International MVJ317
Encompass_Preferred
The Oasis PGA suture with needle is a braided synthetic absorbable suture that is comparable to Ethicon™ Vicryl®. The PGA braided suture has a rapid absorption rate between 60 and 90 days. It retains 60 to 70% of its tensile strength at 14 days. This suture provides excellent pliability and holding strength. Size: 2-0. Needle Type: NSH, 26mm 1/2 Circle Taper Point. Length: 30in. Quantity: 12/box
Catalog No. 50-255-9391