Learn More
Med Vet International Ethicon Plain Gut Suture, Size 4-0, M-1, 12In, Yellowish Tan, 12/Box
Supplier:  Med Vet International 742G
Encompass_Preferred
The Ethicon 742G suture is a high-quality absorbable Plain Gut suture. It measures 12in in length, has a size of 4-0, and are dyed yellowish to provide greater visibility while suturing. This suture comes with a M-1 Reverse Cutting Needle and has 12 sutures in a box.
Catalog No. 50-255-9696