Learn More
Med Vet International Ethicon Prolene Polypropylene Suture, Size 2-0, FSLX, 30In, Blue, 36/Box
Catalog No. 502559623
Encompass_Preferred
The Ethicon 8689H suture is a high-quality non-absorbable Prolene Polypropylene suture. It measures 30in in length, has a size of 2-0, and are dyed blue to provide greater visibility while suturing. This suture comes with a FSLX 36mm 3/8c Reverse Cutting Needle and has 36 sutures in a box.
This item is not returnable. View return policy
Catalog No. 50-255-9623 Supplier Med Vet International Supplier No. 8689H
The Ethicon 8689H suture is a high-quality non-absorbable Prolene Polypropylene suture. It measures 30in in length, has a size of 2-0, and are dyed blue to provide greater visibility while suturing. This suture comes with a FSLX 36mm 3/8c Reverse Cutting Needle and has 36 sutures in a box.