Learn More
Med Vet International Ethicon Prolene Polypropylene Suture, Size 2, LR, 20In, Blue, 24/Box
Supplier:  Med Vet International 3846T
Encompass_Preferred
The Ethicon 3846T suture is a high-quality non-absorbable Prolene Polypropylene suture. It measures 20in in length, has a size of 2, and are dyed blue to provide greater visibility while suturing. This suture comes with a LR 75mm 3/8c Reverse Cutting Needle and has 24 sutures in a box.
Catalog No. 50-255-9719