Learn More
Med Vet International Ethicon Prolene Polypropylene Suture, Size 4-0, RB-1, 30In, Blue, 36/Box
Supplier:  Med Vet International 8871H
Encompass_Preferred
The Ethicon 8871H suture is a high-quality non-absorbable Prolene Polypropylene suture. It measures 30in in length, has a size of 4-0, and are dyed blue to provide greater visibility while suturing. This suture comes with a RB-1 17mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-255-9611
 Restricted Item