Learn More
Med Vet International Dynarex Braided (Pga) Sutures, Absorbable, Synthetic, Violet, 4-0, C6 Needle, 30", 12/Box
Catalog No. 502830276
Encompass_Preferred
Used to hold body tissues together after an injury or surgery. Application generally involves using a needle with an attached length of thread. An absorbable, violet suture made from polyglycolic acid Braided for strength and easy handling and coated to allow for easy passage through tissue High tensile strength during healing period Available in various thread thickness and needle dimensions to accommodate a variety of procedures Sterile
Catalog No. 50-283-0276 Supplier Med Vet International Supplier No. 9130
Used to hold body tissues together after an injury or surgery. Application generally involves using a needle with an attached length of thread. An absorbable, violet suture made from polyglycolic acid Braided for strength and easy handling and coated to allow for easy passage through tissue High tensile strength during healing period Available in various thread thickness and needle dimensions to accommodate a variety of procedures Sterile