Learn More
Med Vet International Dynarex Nylon Sutures, Non Absorbable, Synthetic, Black, 4-0, Pc31 Needle, L-18", 12/Box
Catalog No. 502832226
Encompass_Preferred
Used to hold body tissues together after an injury or surgery. Application generally involves using a needle with an attached length of thread. A non-absorbable suture offering high tensile strength Monofilament for a smooth passage through tissue Available in various thread thickness and needle dimensions to accommodate a variety of procedures Sterile
Catalog No. 50-283-2226 Supplier Med Vet International Supplier No. 9100
Used to hold body tissues together after an injury or surgery. Application generally involves using a needle with an attached length of thread. A non-absorbable suture offering high tensile strength Monofilament for a smooth passage through tissue Available in various thread thickness and needle dimensions to accommodate a variety of procedures Sterile