Learn More
Med Vet International Dynarex Nylon Sutures, Non Absorbable, Synthetic, Black, 5-0, C6 Needle, L-18", 12/Box
Catalog No. 502832232
Encompass_Preferred
Used to hold body tissues together after an injury or surgery. Application generally involves using a needle with an attached length of thread. A non-absorbable suture offering high tensile strength Monofilament for a smooth passage through tissue Available in various thread thickness and needle dimensions to accommodate a variety of procedures Sterile
Catalog No. 50-283-2232 Supplier Med Vet International Supplier No. 9102
Used to hold body tissues together after an injury or surgery. Application generally involves using a needle with an attached length of thread. A non-absorbable suture offering high tensile strength Monofilament for a smooth passage through tissue Available in various thread thickness and needle dimensions to accommodate a variety of procedures Sterile