Learn More
Med Vet International Ethicon Permahand Silk Suture, Size 5-0, FS-2, 18in, 12/Box
Catalog No. 502092861
Encompass_Preferred
The Ethicon 682G suture is a high-quality non-absorbable Silk surgical suture. It measures 18in. in length, has a size of 5-0, and are dyed black to provide greater visibility while suturing. This suture comes with a FS-2 reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-209-2861 Supplier Med Vet International Supplier No. 682G
The Ethicon 682G suture is a high-quality non-absorbable Silk surgical suture. It measures 18in. in length, has a size of 5-0, and are dyed black to provide greater visibility while suturing. This suture comes with a FS-2 reverse cutting needle and has 12 sutures in a box.