Learn More
Med Vet International Ethicon Surgical Steel Suture, Size 4-0, FS-2, 18In, Silver, 12/Box
Supplier:  Med Vet International 603G
Encompass_Preferred
The Ethicon 603G suture is a high-quality non-absorbable Surgical Steel suture. It measures 18in in length, has a size of 4-0, and are dyed silver to provide greater visibility while suturing. This suture comes with a FS-2 19mm 3/8c Reverse Cutting Needle and has 12 sutures in a box.
Catalog No. 50-255-9714