Learn More
Med Vet International Ethicon Vicryl (Polyglactin 910) Suture, Size 1, CT-1, 27in, 36/Box, J341H
Catalog No. 502092867
Encompass_Preferred
The Ethicon J341H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 1, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a CT-1 taper point needle and has 36 sutures in a box.
Catalog No. 50-209-2867 Supplier Med Vet International Supplier No. J341H
The Ethicon J341H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 1, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a CT-1 taper point needle and has 36 sutures in a box.