Learn More
Med Vet International Ethicon Vicryl Suture, Size 2, CP, 27in, 36/Box
Supplier:  Med Vet International J195H
Encompass_Preferred
The Ethicon J195H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 2-0, they undyed and typically have an absorption rate of 56-70 days. This suture comes with a CP-2 reverse cutting needle and has 36 sutures in a box.
Catalog No. 50-255-9596
 Restricted Item