Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Plus Antibacterial Suture, Size 0, CP-1, 27In, Violet, 36/Box
Supplier:  Med Vet International VCP467H
Encompass_Preferred
The Ethicon VCP467H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 27in in length, has a size of 0, and are dyed violet to provide greater visibility while suturing. This suture comes with a CP-1 36mm 1/2c Reverse Cutting Needle and has 36 sutures in a box.
Catalog No. 50-255-9303