Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Plus Antibacterial Suture, Size 0, FSL, 27In, Violet, 36/Box
Supplier:  Med Vet International VCP587H
Encompass_Preferred
The Ethicon VCP587H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 27in in length, has a size of 0, and are dyed violet to provide greater visibility while suturing. This suture comes with a FSL 30mm 3/8c Reverse Cutting Needle and has 36 sutures in a box.
Catalog No. 50-255-9301