Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Suture, Size 3, CCS-1, 27In, Undyed, 24/Box
Supplier:  Med Vet International J808T
Encompass_Preferred
The Ethicon J808T suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 27in in length, has a size of 3, and are undyed. This suture comes with a Conventional Cutting Needle and has 24 sutures in a box.
Catalog No. 50-255-9570
 Restricted Item