Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Suture, Size 5-0, PS-5, 18In, Undyed, 12/Box
Supplier:  Med Vet International J593G
Encompass_Preferred
The Ethicon J593G suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 18in in length, has a size of 5-0, and are undyed. This suture comes with a Reverse Prime Cutting Needle and has 12 sutures in a box.
Catalog No. 50-255-9573