Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Suture, Size 8-0, Double Armed TG 140-8 TG 140-8, 8In, Violet, 12/Box
Supplier:  Med Vet International J547G
Encompass_Preferred
The Ethicon J547G suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 8in in length, has a size of 8-0, and are dyed violet to provide greater visibility while suturing. This suture comes with a Double Armed TG 140-8 TG 140-8 6.5mm 3/8c Spatula Needle and has 12 sutures in a box.
Catalog No. 50-255-9578