Learn More
Med Vet International Ethicon Vicryl Rapide Suture, Size 5-0, P-1, 18In, Undyed, 12/Box
Supplier:  Med Vet International VR490
Encompass_Preferred
The Ethicon VR490 suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 18in in length, has a size of 5-0, and are undyed. This suture comes with a P-1 Precision Point Reverse Cutting Needle and has 12 sutures in a box.
Catalog No. 50-255-9298