Learn More
Med Vet International Ethicon Coated Vicryl Plus Antibacterial Suture, Size 0, CT-2, 27in, 36/Box
Catalog No. 502092823
Encompass_Preferred
The Ethicon VCP334H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a CT-2 taper point needle and has 36 sutures in a box. Primary code substitute for discontinued code J334H
Catalog No. 50-209-2823 Supplier Med Vet International Supplier No. VCP334H
The Ethicon VCP334H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a CT-2 taper point needle and has 36 sutures in a box. Primary code substitute for discontinued code J334H