Learn More
Med Vet International Ethicon Coated Vicryl Plus Antibacterial Suture, Size 3-0, SH-1, 27in, 36/Box
Catalog No. 502092828
Encompass_Preferred
The Ethicon VCP311H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 3-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a SH-1 taper point needle and has 36 sutures in a box. Primary code substitute for discontinued code J311H
Catalog No. 50-209-2828 Supplier Med Vet International Supplier No. VCP311H
 Restricted Item
Add to Cart
Add to Cart
The Ethicon VCP311H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 3-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a SH-1 taper point needle and has 36 sutures in a box. Primary code substitute for discontinued code J311H