Learn More
Med Vet International Ethicon Coated Vicryl Plus Antibacterial Suture, Size 4-0, FS-2, 27in, 36/Box
Catalog No. 502092829
Encompass_Preferred
The Ethicon VCP422H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 4-0, they are dyed Violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a FS-2 reverse cutting needle and has 36 sutures in a box. Primary code substitute for discontinued code J422H
Catalog No. 50-209-2829 Supplier Med Vet International Supplier No. VCP422H
The Ethicon VCP422H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 4-0, they are dyed Violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a FS-2 reverse cutting needle and has 36 sutures in a box. Primary code substitute for discontinued code J422H