Learn More
Med Vet International Ethicon Vicryl Suture, Size 5-0, P-3, 18in, Violet, 12/Box
Catalog No. 502092818
Encompass_Preferred
The Ethicon J463G suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 5-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-209-2818 Supplier Med Vet International Supplier No. J463G
The Ethicon J463G suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 5-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.