Learn More
Med Vet International Ethicon Vicryl Suture, Size 9-0, TG140-8, 12in, 12/Box
Catalog No. 502092821
Encompass_Preferred
The Ethicon V549G suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 12in. in length, has a size of 9-0, they are dyed black to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a TG140-8 Spatula reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-209-2821 Supplier Med Vet International Supplier No. V549G
The Ethicon V549G suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 12in. in length, has a size of 9-0, they are dyed black to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a TG140-8 Spatula reverse cutting needle and has 12 sutures in a box.