Learn More
Eurofins Discoverx SUV39H2, 50 µg. (EPI065)
Supplier:  Eurofins Discoverx EPI065
Encompass
SUV39H2 is purified, untagged recombinant SUV39H2 enzyme. This enzyme contains amino acids 2-248. It retains its binding/enzymatic activity and is intended for use in epigenetic assays. This enzyme is provided as 1 mg/mL frozen in 50% glycerol. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-200-5293