Learn More
Aviva Systems Biology Corp SUV39H2 Antibody - middle region (ARP48910_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48910_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUV39H2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SUV39H2. Alternate names: FLJ23414, KMT1BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-388