Learn More
Aviva Systems Biology Corp KMT5B Antibody - C-terminal region (ARP34567_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34567_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SUV420H1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SUV420H1. Alternate names: CGI85, KMT5B, CGI-85ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-52967