Learn More
Aviva Systems Biology Corp KMT5B Antibody - C-terminal region (ARP51247_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51247_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Suv420h1. It was validated on Western Blot. . Alternate names: AA117471, C630029K18Rik, KMT5B, MGC18702, Suv4-20h1ApplicationCHIP, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-567