Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Suvorexant impurity 2 | 1149352-55-5 | 99.7% | C10H9N3O2 | 50 MG
SDP

Catalog No. 5000433947
Encompass_Preferred

Suvorexant impurity 2 is a chemical impurity of Suvorexant, presented as a solid. It is intended for research purposes only.

  • High purity: 99.7%
  • Chemical formula: C10H9N3O2
  • CAS number: 1149352-55-5
  • Appearance: solid
  • Recommended storage at room temperature for 3 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
  • Shipping at room temperature

Catalog No. 50-004-33947 Supplier Medchemexpress LLC Supplier No. HY7700250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suvorexant impurity 2 is a chemical impurity of Suvorexant, presented as a solid. It is intended for research purposes only.

  • High purity: 99.7%
  • Chemical formula: C10H9N3O2
  • CAS number: 1149352-55-5
  • Appearance: solid
  • Recommended storage at room temperature for 3 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
  • Shipping at room temperature

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.