Learn More
Aviva Systems Biology Corp SUZ12 Antibody - middle region (ARP32515_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32515_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUZ12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SUZ12. Alternate names: CHET9, JJAZ1, KIAA0160ApplicationCHIP, IHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54629