Learn More
Cayman Chemical Suzetrgn 10mg
SDP
Catalog No. 502943464
Encompass_Preferred
A Nav1.8 inhibitor (IC50 = 0.7 nM for the human channel); selective for Nav1.8 over a panel of eight additional voltage-gated sodium channels (IC50s = >10,000 nM for all)
Catalog No. 50-294-3464 Supplier Cayman Chemical Supplier No. 4079910 MG
A Nav1.8 inhibitor (IC50 = 0.7 nM for the human channel); selective for Nav1.8 over a panel of eight additional voltage-gated sodium channels (IC50s = >10,000 nM for all)