Learn More
Medchemexpress LLC Suzetrigine phenol | 2649467-91-2 | 99.4% | 459.37 | 1 MG
SDP
Catalog No. 5000392256
Encompass_Preferred
Suzetrigine phenol is the phenolate form of the sodium channel modulator Suzetrigine. It is an orally active and highly selective inhibitor of NaV1.8.
  • Phenolate form of Suzetrigine
  • Orally active and highly selective inhibitor of NaV1.8
  • For research use only
  • Modulates sodium channels
Catalog No. 50-003-92256 Supplier Medchemexpress LLC Supplier No. HY1609031MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Suzetrigine phenol is the phenolate form of the sodium channel modulator Suzetrigine. It is an orally active and highly selective inhibitor of NaV1.8.
  • Phenolate form of Suzetrigine
  • Orally active and highly selective inhibitor of NaV1.8
  • For research use only
  • Modulates sodium channels