Learn More
Medchemexpress LLC Suzetrigine phenol | 2649467-91-2 | 99.4% | 459.37 | 25 MG
SDP
Catalog No. 5000391893
Encompass_Preferred
Suzetrigine phenol is the phenolate form of the sodium channel modulator Suzetrigine. Suzetrigine is an orally active and highly selective inhibitor of NaV1.8, making it suitable for research into neurological and eye/ear diseases.
  • Phenolate form
  • Orally active and highly selective inhibitor of NaV1.8
  • Supports neurological disease research
  • Supports eye or ear disease research
Catalog No. 50-003-91893 Supplier Medchemexpress LLC Supplier No. 25MGHY160903
Suzetrigine phenol is the phenolate form of the sodium channel modulator Suzetrigine. Suzetrigine is an orally active and highly selective inhibitor of NaV1.8, making it suitable for research into neurological and eye/ear diseases.
  • Phenolate form
  • Orally active and highly selective inhibitor of NaV1.8
  • Supports neurological disease research
  • Supports eye or ear disease research