Learn More
Medchemexpress LLC Suzetrigine phenol | 2649467-91-2 | 99.4% | 459.37 | 5 MG
SDP
Catalog No. 5000393034
Encompass_Preferred
Suzetrigine phenol is the phenolate form of the sodium channel modulator Suzetrigine (HY-148800). Suzetrigine (VX-548) is an orally active and highly selective inhibitor of NaV1.8.
  • Phenolate form of sodium channel modulator
  • Orally active
  • Highly selective inhibitor of NaV1.8
Catalog No. 50-003-93034 Supplier Medchemexpress LLC Supplier No. HY1609035MG
Suzetrigine phenol is the phenolate form of the sodium channel modulator Suzetrigine (HY-148800). Suzetrigine (VX-548) is an orally active and highly selective inhibitor of NaV1.8.
  • Phenolate form of sodium channel modulator
  • Orally active
  • Highly selective inhibitor of NaV1.8