Learn More
Azzota Corp SV1800 Vis Spectro 320-1100nm
Catalog No. 5000887263
Encompass_Preferred
SV1800 Visible Spectrophotometer 2nm bandwidth 320-1100nm 128x64 LCD Automatically set WL USB connection Can display and save 200 standard curves and 200 groups of data With Kinetics Function
Catalog No. 50-008-87263 Supplier Azzota Corp Supplier No. SV1800
SV1800 Visible Spectrophotometer 2nm bandwidth 320-1100nm 128x64 LCD Automatically set WL USB connection Can display and save 200 standard curves and 200 groups of data With Kinetics Function